Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032777.9 | NP_116166.9 | 19 | 5389 | ||
XM_011544481.2 | XP_011542783.1 | 19 | 5458 | |||
XM_011544482.2 | XP_011542784.1 | 18 | 5386 | |||
XM_011544483.2 | XP_011542785.1 | 18 | 5299 | |||
GenBank mRNA | AB040964.2 | - | 16 | 4715 | ||
AF378755.1 | - | 19 | 5016 | |||
AK027296.1 | - | 6 | 1429 | |||
AK074099.2 | - | 19 | 4920 | |||
AL110244.1 | - | 1 | 2010 | |||
AL137610.1 | - | 1 | 488 | |||
AL832524.1 | - | 3 | 2456 | |||
AL832959.1 | - | 12 | 4059 | |||
BC146774.1 | - | 16 | 4715 | 9007223 |
Set Membership: |
![]() |