Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CECR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017829.5 | NP_060299.4 | 8 | 903 | ||
NM_033070.2 | NP_149061.1 | 8 | 978 | |||
XM_005261236.3 | XP_005261293.1 | 8 | 956 | |||
XM_011546126.2 | XP_011544428.1 | 7 | 774 | |||
XM_011546127.1 | XP_011544429.1 | 8 | 1393 | |||
XM_017028781.1 | XP_016884270.1 | 8 | 1445 | |||
XM_017028782.1 | XP_016884271.1 | 9 | 1772 | |||
XM_017028783.1 | XP_016884272.1 | 9 | 2209 | |||
XM_017028784.1 | XP_016884273.1 | 9 | 1772 | |||
GenBank mRNA | AF273270.1 | - | 8 | 903 | ||
AF273271.1 | - | 8 | 963 | |||
AK000461.1 | - | 8 | 976 | |||
AK001034.1 | - | 8 | 956 | |||
AK123646.1 | - | 3 | 1459 | |||
AK315156.1 | - | 8 | 964 | |||
AL833450.1 | - | 3 | 2794 | |||
BC042540.1 | - | 8 | 962 | 4342582 | ||
BM558767.1 | - | 7 | 788 |