Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085347.2 | NP_001078816.1 | 2 | 377 | ||
NM_001134430.2 | NP_001127902.1 | 2 | 377 | |||
NM_130459.3 | NP_569726.2 | 2 | 377 | |||
XM_011518554.1 | XP_011516856.1 | 2 | 415 | |||
GenBank mRNA | AK027677.1 | - | 2 | 312 | ||
AK075520.1 | - | 2 | 320 | |||
AK092388.1 | - | 1 | 890 | |||
AY358180.1 | - | 2 | 344 | |||
CN263138.1 | - | 2 | 379 | |||
DA430836.1 | - | 2 | 353 |