Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013371.3 | NP_037503.2 | 7 | 1397 | ||
NM_153758.2 | NP_715639.1 | 6 | 597 | |||
GenBank mRNA | AF192498.1 | - | 6 | 505 | ||
AF453946.1 | - | 7 | 1397 | |||
AY040367.1 | - | 6 | 597 |