Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039592.1 | NP_001034681.1 | 5 | 1241 | ||
NM_172312.1 | NP_758516.1 | 5 | 1241 | |||
GenBank mRNA | AK302313.1 | - | 5 | 1137 | ||
BC019247.1 | - | 5 | 1146 | 4825175 | ||
S83157.1 | - | 5 | 1242 | |||
U12978.2 | - | 5 | 1242 |