Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022075.4 | NP_071358.1 | 8 | 1085 | ||
NM_181746.3 | NP_859530.1 | 8 | 1125 | |||
XM_011509451.2 | XP_011507753.1 | 8 | 788 | |||
XM_011509452.2 | XP_011507754.1 | 8 | 836 | |||
GenBank mRNA | AF177338.2 | - | 8 | 814 | ||
AF189062.4 | - | 3 | 298 | |||
AK001105.1 | - | 7 | 754 | |||
AK290989.1 | - | 8 | 761 | |||
AY091458.1 | - | 8 | 907 | |||
BC001357.2 | - | 8 | 780 | 3051086 | ||
BC010032.2 | - | 8 | 744 | 3506408 |