Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005829.4 | NP_005820.1 | 6 | 1351 | ||
NR_023361.1 | 7 | 1470 | ||||
NR_037582.1 | 6 | 1347 | ||||
GenBank mRNA | AK294712.1 | - | 7 | 1062 | ||
BC002785.2 | - | 6 | 936 | 3614033 | ||
BC007773.2 | - | 6 | 935 | |||
BC010020.2 | - | 6 | 963 | 2959670 | ||
BX098014.1 | - | 1 | 27 | |||
X99459.1 | - | 6 | 945 |
Gene Symbol: C15orf38-AP3S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199058.1 | NP_001185987.1 | 10 | 1673 | ||
GenBank mRNA | AK000495.1 | - | 10 | 1565 |