Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003759.3 | NP_003750.1 | 1 | 281 | ||
GenBank mRNA | AF007216.1 | - | 1 | 234 | ||
BC030977.1 | - | 1 | 281 | 4610968 | ||
DA635432.1 | - | 1 | 290 |