Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005870.4 | NP_005861.2 | 4 | 2001 | ||
GenBank mRNA | BC030836.1 | - | 4 | 1970 | 4794888 |