Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TNIP1 SIRNA Assays ›
Gene Symbol:
TNIP1
Gene Name:
TNFAIP3 interacting protein 1
Gene Aliases:
ABIN-1, NAF1, VAN, nip40-1
Chromosome Location:
Chr.5: 151029943 - 151087660 on Build GRCh38
UniGene ID:
Hs.355141
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
135564
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TNIP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001252385.1
NP_001239314.1
6
840
NM_001252386.1
NP_001239315.1
5
668
NM_001252390.1
NP_001239319.1
6
705
NM_001252391.1
NP_001239320.1
6
737
NM_001252392.1
NP_001239321.1
6
737
NM_001252393.1
NP_001239322.1
6
840
NM_001258454.1
NP_001245383.1
6
840
NM_001258455.1
NP_001245384.1
5
635
NM_001258456.1
NP_001245385.1
5
635
NM_006058.4
NP_006049.3
6
840
XM_005268355.1
XP_005268412.1
6
753
XM_006714751.1
XP_006714814.1
6
753
XM_006714752.2
XP_006714815.1
6
753
XM_017008945.1
XP_016864434.1
6
753
XM_017008946.1
XP_016864435.1
6
719
XM_017008947.1
XP_016864436.1
7
1222
XM_017008948.1
XP_016864437.1
5
1014
XM_017008949.1
XP_016864438.1
6
753
XM_017008950.1
XP_016864439.1
6
753
XM_017008951.1
XP_016864440.1
5
1014
GenBank mRNA
AB177543.1
-
5
599
AB177544.1
-
5
599
AB252970.1
-
6
709
AB252971.1
-
6
709
AB252972.1
-
6
830
AB252973.1
-
6
830
AB252976.1
-
4
463
AB252977.1
-
4
463
AB252978.1
-
4
463
AB252979.1
-
4
463
AB252980.1
-
4
463
AB252981.1
-
4
463
AB252982.1
-
6
709
AJ011895.1
-
6
709
AJ011896.1
-
6
709
AK074400.1
-
6
771
AK290999.1
-
6
706
AK298685.1
-
5
662
AK299975.1
-
5
600
AK303573.1
-
6
790
AY012155.1
-
6
856
BC012133.1
-
6
746
4549542
BC014008.1
-
6
687
4040118
BX640647.1
-
5
589
D30755.2
-
5
599
DA643531.1
-
4
320
U39403.1
-
2
195
U83844.1
-
3
320
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
MyD88-dependent toll-like receptor signaling pathway
translation
proteolysis
defense response
inflammatory response
leukocyte cell-cell adhesion
glycoprotein biosynthetic process
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of viral genome replication
positive regulation of transcription from RNA polymerase II promoter
positive regulation of inflammatory response
negative regulation of ERK1 and ERK2 cascade
modulation by symbiont of host I-kappaB kinase/NF-kappaB cascade
thiol-dependent ubiquitin-specific protease activity
protein binding
mitogen-activated protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-kprhh:80/10.239.160.57:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline