Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRVI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098579.2 | NP_001092049.2 | 18, 19 | 2421 | ||
NM_001100163.2 | NP_001093633.1 | 19, 20 | 2699 | |||
NM_001100167.2 | NP_001093637.1 | 19, 20 | 2416 | |||
NM_001206880.1 | NP_001193809.1 | 18, 19 | 2134 | |||
NM_001206881.1 | NP_001193810.1 | 17, 18 | 2541 | |||
NM_130385.3 | NP_569056.4 | 19, 20 | 2752 | |||
GenBank mRNA | AF081249.1 | - | 17, 18 | 2437 | ||
AF081250.1 | - | 17, 18 | 2165 | |||
AK127209.1 | - | 19, 20 | 2394 | |||
AK296336.1 | - | 19, 20 | 2516 | |||
AK300358.1 | - | 18, 19 | 2406 | |||
AK304702.1 | - | 18, 19 | 1768 | |||
BC117127.1 | - | 19, 20 | 2289 | 40125678 |