Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006108.3 | NP_006099.2 | 16 | 3411 | ||
GenBank mRNA | AB018305.2 | - | 15 | 2896 | ||
AK001584.1 | - | 8 | 1980 | |||
AK074803.1 | - | 16 | 3357 | |||
BC019825.1 | - | 5 | 1501 | |||
BC041974.1 | - | 4 | 1494 |