Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006351.3 | NP_006342.2 | 7, 8 | 948 | ||
GenBank mRNA | AF026030.1 | - | 7, 8 | 765 | ||
AF041254.1 | - | 7, 8 | 806 | |||
AK223062.1 | - | 7, 8 | 791 | |||
AK289634.1 | - | 7, 8 | 948 | |||
AK309415.1 | - | 8, 9 | 899 | |||
BC033628.1 | - | 7, 8 | 778 | 5248163 |