Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105530.1 | NP_001099000.1 | 13 | 2340 | ||
NM_006367.3 | NP_006358.1 | 13 | 2358 | |||
XM_005270368.1 | XP_005270425.1 | 13 | 2198 | |||
XM_011540509.1 | XP_011538811.1 | 14 | 2751 | |||
XM_011540510.1 | XP_011538812.1 | 14 | 2748 | |||
XM_011540511.1 | XP_011538813.1 | 13 | 2182 | |||
XM_011540512.1 | XP_011538814.1 | 13 | 2301 | |||
XM_011540513.2 | XP_011538815.1 | 13 | 2193 | |||
XM_011540514.1 | XP_011538816.1 | 14 | 2308 | |||
XM_011540515.1 | XP_011538817.1 | 14 | 2748 | |||
XM_017000067.1 | XP_016855556.1 | 13 | 2266 | |||
XM_017000068.1 | XP_016855557.1 | 14 | 2310 | |||
XM_017000069.1 | XP_016855558.1 | 14 | 2521 | |||
XM_017000070.1 | XP_016855559.1 | 13 | 2540 | |||
XM_017000071.1 | XP_016855560.1 | 13 | 2498 | |||
XM_017000072.1 | XP_016855561.1 | 13 | 2434 | |||
XM_017000073.1 | XP_016855562.1 | 13 | 2255 | |||
GenBank mRNA | AK096193.1 | - | 13 | 2192 | ||
BC013963.2 | - | 13 | 2175 | 2963922 | ||
BC020367.1 | - | 2 | 897 | |||
BM772471.1 | - | 1 | 585 | |||
L12168.1 | - | 12 | 2168 |