Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271977.1 | NP_001258906.1 | 12 | 1644 | ||
NM_001271978.1 | NP_001258907.1 | 11 | 1616 | |||
NM_001271979.1 | NP_001258908.1 | 11 | 1413 | |||
NM_001271980.1 | NP_001258909.1 | 11 | 1351 | |||
NM_001282305.1 | NP_001269234.1 | 11 | 1611 | |||
NM_001282306.1 | NP_001269235.1 | 12 | 1594 | |||
NM_001282307.1 | NP_001269236.1 | 11 | 1514 | |||
NM_001282308.1 | NP_001269237.1 | 11 | 1452 | |||
NM_006374.4 | NP_006365.2 | 12 | 1582 | |||
XM_011510494.2 | XP_011508796.1 | 12 | 1447 | |||
XM_011510495.2 | XP_011508797.1 | 11 | 1719 | |||
XM_017003168.1 | XP_016858657.1 | 11 | 1521 | |||
XM_017003169.1 | XP_016858658.1 | 11 | 1402 | |||
XM_017003170.1 | XP_016858659.1 | 11 | 1350 | |||
XM_017003171.1 | XP_016858660.1 | 10 | 1322 | |||
GenBank mRNA | AK092161.1 | - | 11 | 1274 | ||
AK291808.1 | - | 11 | 1240 | |||
AK291947.1 | - | 12 | 1471 | |||
AK297374.1 | - | 11 | 1403 | |||
AK301214.1 | - | 12 | 1482 | |||
AK303719.1 | - | 11 | 1584 | |||
AK315966.1 | - | 11 | 1352 | |||
BC007852.2 | - | 12 | 1506 | 4298239 | ||
BC015793.1 | - | 11 | 1562 | 4867978 | ||
BC091505.1 | - | 12 | 1536 | 30383354 | ||
CR936874.1 | - | 2 | 4307 | |||
D63780.1 | - | 11 | 1405 | |||
X99325.1 | - | 11 | 1417 |