Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006391.2 | NP_006382.1 | 1, 2 | 225 | ||
GenBank mRNA | AF098799.1 | - | 1, 2 | 190 | ||
AK312885.1 | - | 1, 2 | 196 | |||
BC114929.1 | - | 1, 2 | 151 | 40036258 | ||
DA780921.1 | - | 1, 2 | 225 |