Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300929.1 | NP_001287858.1 | 3 | 594 | ||
NM_003795.5 | NP_003786.1 | 3 | 660 | |||
NM_152827.3 | NP_690040.1 | 2 | 564 | |||
GenBank mRNA | AB047360.1 | - | 2 | 268 | ||
AF034546.1 | - | 3 | 364 | |||
AF062483.1 | - | 3 | 340 | |||
AK289476.1 | - | 3 | 451 | |||
BC008444.1 | - | 3 | 418 | 4102735 | ||
BC014580.1 | - | 3 | 442 | 3932208 | ||
BC015179.1 | - | 3 | 424 | 3905515 | ||
BC016863.2 | - | 4 | 761 | 3846511 | ||
BT007114.1 | - | 3 | 321 | |||
CN422502.1 | - | 3 | 340 | |||
CR456898.1 | - | 3 | 321 |