Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252641.1 | NP_001239570.1 | 4 | 264 | ||
NM_003796.3 | NP_003787.2 | 4 | 579 | |||
NR_045557.1 | 3 | 544 | ||||
XM_005259362.2 | XP_005259419.1 | 4 | 258 | |||
XM_005259363.4 | XP_005259420.1 | 4 | 315 | |||
XM_011527435.1 | XP_011525737.1 | 3 | 141 | |||
GenBank mRNA | AF091095.1 | - | 5 | 215 | ||
AK001437.1 | - | 3 | 265 | |||
AK226173.1 | - | 4 | 567 | |||
AK292170.1 | - | 4 | 315 | |||
AK314313.1 | - | 4 | 304 | |||
BC026184.2 | - | 4 | 264 | |||
BC067259.1 | - | 3 | 271 | 6500727 |