Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001248006.1 | NP_001234935.1 | 3, 4 | 768 | ||
NM_001248007.1 | NP_001234936.1 | 2, 3 | 431 | |||
NM_006458.3 | NP_006449.2 | 4, 5 | 769 | |||
NM_033278.3 | NP_150594.2 | 3, 4 | 604 | |||
XM_011519844.1 | XP_011518146.1 | 4, 5 | 868 | |||
XM_011519845.2 | XP_011518147.1 | 3, 4 | 549 | |||
XM_017017098.1 | XP_016872587.1 | 3, 4 | 580 | |||
XM_017017099.1 | XP_016872588.1 | 3, 4 | 472 | |||
GenBank mRNA | AF045239.1 | - | 2, 3 | 373 | ||
AF220020.1 | - | 2, 3 | 373 | |||
AF220021.1 | - | 2, 3 | 373 | |||
AF220022.1 | - | 2, 3 | 373 | |||
AK291396.1 | - | 3, 4 | 768 | |||
AK298843.1 | - | 2, 3 | 307 | |||
AK299586.1 | - | 3, 4 | 561 | |||
AK311127.1 | - | 3, 4 | 559 | |||
AL360192.1 | - | 2, 3 | 343 | |||
BC096827.1 | - | 3, 4 | 604 | 6108991 | ||
BM758857.1 | - | 1, 2 | 56 | |||
BU500178.1 | - | 614 | ||||
DA089323.1 | - | 3, 4 | 479 |