Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303618.1 | NP_001290547.1 | 2 | 670 | ||
NM_006566.3 | NP_006557.2 | 3 | 672 | |||
XM_005266642.3 | XP_005266699.1 | 3 | 771 | |||
XM_017025524.1 | XP_016881013.1 | 2 | 670 | |||
GenBank mRNA | AJ537553.1 | - | 2 | 252 | ||
AK313199.1 | - | 2 | 671 | |||
BC074787.2 | - | 2 | 257 | 30915562 | ||
GU935335.1 | - | 3 | 419 | |||
U56102.1 | - | 3 | 431 |