Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006576.3 | NP_006567.3 | 7 | 859 | ||
XM_017018710.1 | XP_016874199.1 | 8 | 925 | |||
XM_017018711.1 | XP_016874200.1 | 8 | 3671 | |||
XM_017018712.1 | XP_016874201.1 | 8 | 3614 | |||
XM_017018713.1 | XP_016874202.1 | 8 | 3274 | |||
GenBank mRNA | AF041449.1 | - | 7 | 859 | ||
AK314362.1 | - | 7 | 1261 | |||
BC111730.1 | - | 8 | 963 | 40035028 |