Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006583.2 | NP_006574.1 | 5 | 680 | ||
GenBank mRNA | AF012270.1 | - | 5 | 696 | ||
BC128120.1 | - | 5 | 671 | 40112275 | ||
BC128121.1 | - | 5 | 671 | 40112276 | ||
BC128401.1 | - | 5 | 671 | 40112297 |
Set Membership: |
![]() |