Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256725.1 | NP_001243654.1 | 11, 12 | 1663 | ||
NM_001256727.1 | NP_001243656.1 | 11, 12 | 1548 | |||
NM_001256728.1 | NP_001243657.1 | 10, 11 | 1473 | |||
NM_006590.3 | NP_006581.2 | 11, 12 | 1663 | |||
NR_046347.1 | 11, 12 | 1551 | ||||
XM_006711922.1 | XP_006711985.1 | 11, 12 | 1531 | |||
XM_006711923.2 | XP_006711986.1 | 11, 12 | 1610 | |||
XM_011532487.1 | XP_011530789.1 | 11, 12 | 1616 | |||
XM_011532488.1 | XP_011530790.1 | 11, 12 | 1549 | |||
XM_017003182.1 | XP_016858671.1 | 11, 12 | 1620 | |||
GenBank mRNA | AF130096.1 | - | 11, 12 | 1252 | ||
AF132955.1 | - | 11, 12 | 1560 | |||
AF161450.1 | - | 6, 7 | 649 | |||
AF353989.1 | - | 11, 12 | 1570 | |||
AK001772.1 | - | 7, 8 | 956 | |||
AK056334.1 | - | 11, 12 | 1532 | |||
AK129872.1 | - | 12, 13 | 1544 | |||
AK222852.1 | - | 11, 12 | 1566 | |||
AK289451.1 | - | 11, 12 | 1617 | |||
AK295257.1 | - | 10, 11 | 1471 | |||
AK302236.1 | - | 11, 12 | 1525 | |||
AL117416.1 | - | 9, 10 | 1230 | |||
BC001384.2 | - | 11, 12 | 1552 | |||
BC067273.1 | - | 11, 12 | 1591 | 6503509 |