Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEPT9 SIRNA Assays ›
Gene Symbol:
SEPT9
Gene Name:
septin 9
Gene Aliases:
AF17q25, MSF, MSF1, NAPB, PNUTL4, SINT1, SeptD1
Chromosome Location:
Chr.17: 77281410 - 77500596 on Build GRCh38
UniGene ID:
Hs.440932
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
135964
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEPT9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113491.1
NP_001106963.1
12
2196
NM_001113492.1
NP_001106964.1
12
2371
NM_001113493.1
NP_001106965.1
11
2357
NM_001113494.1
NP_001106966.1
11
2310
NM_001113495.1
NP_001106967.1
10
2010
NM_001113496.1
NP_001106968.1
10
1605
NM_001293695.1
NP_001280624.1
11
2139
NM_001293696.1
NP_001280625.1
10
1421
NM_001293697.1
NP_001280626.1
10
1634
NM_001293698.1
NP_001280627.1
10
1720
NM_006640.4
NP_006631.2
11
2828
XM_005256962.1
XP_005257019.1
10
1478
XM_006721643.2
XP_006721706.1
13
2299
XM_006721644.1
XP_006721707.1
10
1521
XM_011524204.1
XP_011522506.1
13
2396
XM_011524206.1
XP_011522508.1
11
2116
XM_011524207.1
XP_011522509.1
11
2129
XM_011524208.2
XP_011522510.1
10
1387
XM_017024031.1
XP_016879520.1
11
1728
XM_017024032.1
XP_016879521.1
10
1727
GenBank mRNA
AB023208.1
-
11
2311
AF142408.1
-
11
2273
AF142569.1
-
1
227
AF189712.2
-
13
2324
AF189713.3
-
12
2167
AJ312319.1
-
11
2358
AJ312320.1
-
10
1400
AK056495.1
-
10
2011
AK299828.1
-
10
1635
AK300270.1
-
11
2099
AK316473.1
-
10
1721
AL080131.1
-
7
1016
BC021192.2
-
11
2052
3346534
BC054004.1
-
13
2027
6146448
DB007851.1
-
2
498
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cell cycle
protein heterooligomerization
cell division
cell-cell adhesion
positive regulation of nonmotile primary cilium assembly
GTPase activity
protein binding
GTP binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-cs4xr:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0