Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBGCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256617.1 | NP_001243546.1 | 8 | 1371 | ||
NM_001256618.1 | NP_001243547.1 | 6 | 1098 | |||
NM_006659.3 | NP_006650.1 | 7 | 1287 | |||
NR_046330.1 | 7 | 1671 | ||||
GenBank mRNA | AF042379.1 | - | 7 | 977 | ||
AK092113.1 | - | 7 | 1671 | |||
AK223586.1 | - | 7 | 1178 | |||
AK297251.1 | - | 6 | 820 | |||
AK299564.1 | - | 7 | 841 | |||
AK314520.1 | - | 7 | 1009 | |||
BC093770.1 | - | 6 | 966 | 7939615 | ||
BC111957.1 | - | 6 | 966 | 8327425 | ||
BC143247.1 | - | 7 | 1050 | 9051749 |