Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEFF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003692.4 | NP_003683.2 | 2 | 524 | ||
GenBank mRNA | BC035056.1 | - | 2 | 326 | 5262325 | |
DN602026.1 | - | 2 | 524 |
Gene Symbol: MSANTD3-TMEFF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198812.1 | NP_001185741.1 | 2 | 472 | ||
GenBank mRNA | AL831919.1 | - | 2 | 107 |