Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130912.2 | NP_001124384.1 | 11 | 939 | ||
NM_001320831.1 | NP_001307760.1 | 11 | 971 | |||
NM_006704.4 | NP_006695.1 | 10 | 843 | |||
XM_011534891.2 | XP_011533193.1 | 11 | 1034 | |||
XM_011534893.2 | XP_011533195.1 | 10 | 752 | |||
XM_017020365.1 | XP_016875854.1 | 11 | 924 | |||
XM_017020366.1 | XP_016875855.1 | 11 | 799 | |||
XM_017020367.1 | XP_016875856.1 | 11 | 833 | |||
XM_017020368.1 | XP_016875857.1 | 11 | 845 | |||
GenBank mRNA | AF132856.1 | - | 10 | 613 | ||
AJ344097.1 | - | 10 | 706 | |||
AK291402.1 | - | 11 | 756 | |||
AK292128.1 | - | 10 | 703 | |||
AK302363.1 | - | 11 | 818 | |||
AY271314.1 | - | 11 | 776 | |||
AY321358.1 | - | 11 | 791 | |||
BC000911.1 | - | 10 | 668 | 2985858 | ||
BT009798.1 | - | 10 | 613 |