Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286625.1 | NP_001273554.1 | 6 | 898 | ||
NM_001286626.1 | NP_001273555.1 | 5 | 905 | |||
NM_001286627.1 | NP_001273556.1 | 5 | 905 | |||
NM_006711.4 | NP_006702.1 | 6 | 959 | |||
NM_080594.3 | NP_542161.1 | 6 | 1093 | |||
NR_104485.1 | 5 | 1027 | ||||
XM_005255048.1 | XP_005255105.1 | 6 | 1186 | |||
XM_005255049.3 | XP_005255106.1 | 6 | 2651 | |||
XM_017022874.1 | XP_016878363.1 | 6 | 1552 | |||
GenBank mRNA | AB445101.1 | - | 5 | 618 | ||
AF015608.1 | - | 5 | 632 | |||
AF247662.1 | - | 3 | 387 | |||
AF274003.1 | - | 5 | 594 | |||
AK289955.1 | - | 7 | 853 | |||
AK293343.1 | - | 5 | 610 | |||
AK303100.1 | - | 5 | 566 | |||
AK316132.1 | - | 5 | 895 | |||
AL559988.3 | - | 5 | 679 | |||
BC001659.2 | - | 6 | 783 | 2959954 | ||
BC001838.2 | - | 6 | 783 | 2959954 | ||
BC108316.1 | - | 6 | 904 | 6151864 | ||
BI819738.1 | - | 5 | 589 | |||
BQ059026.1 | - | 2 | 761 | |||
L37368.1 | - | 6 | 1167 |