Search Thermo Fisher Scientific
Gene Symbol: MALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006785.3 | NP_006776.1 | 17 | 4691 | ||
NM_173844.2 | NP_776216.1 | 16 | 4658 | |||
GenBank mRNA | AF070528.1 | - | 1 | 1230 | ||
AL137399.1 | - | 3 | 2643 |