Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007217.3 | NP_009148.2 | 9 | 1281 | ||
NM_145859.1 | NP_665858.1 | 9 | 1130 | |||
NM_145860.1 | NP_665859.1 | 8 | 1036 | |||
XM_005247086.4 | XP_005247143.1 | 9 | 2016 | |||
XM_005247087.4 | XP_005247144.1 | 9 | 2021 | |||
XM_005247088.3 | XP_005247145.1 | 8 | 1112 | |||
XM_006713485.3 | XP_006713548.1 | 9 | 1176 | |||
XM_011512368.2 | XP_011510670.1 | 10 | 2089 | |||
XM_011512369.2 | XP_011510671.1 | 10 | 2094 | |||
XM_017005644.1 | XP_016861133.1 | 9 | 1171 | |||
XM_017005645.1 | XP_016861134.1 | 9 | 2012 | |||
GenBank mRNA | AF022385.1 | - | 8 | 1037 | ||
BC002506.1 | - | 9 | 1293 | 3050540 | ||
BC016353.1 | - | 9 | 1134 | 4092244 |