Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161521.1 | NP_001154993.1 | 5 | 541 | ||
NM_001161522.1 | NP_001154994.1 | 4 | 434 | |||
NM_007246.3 | NP_009177.3 | 5 | 664 | |||
XM_011531572.2 | XP_011529874.1 | 6 | 654 | |||
XM_011531575.2 | XP_011529877.1 | 4 | 486 | |||
XM_011531576.2 | XP_011529878.1 | 3 | 154 | |||
XM_017007674.1 | XP_016863163.1 | 6 | 614 | |||
XM_017007675.1 | XP_016863164.1 | 4 | 379 | |||
XM_017007676.1 | XP_016863165.1 | 4 | 487 | |||
XM_017007677.1 | XP_016863166.1 | 3 | 361 | |||
GenBank mRNA | AF059569.1 | - | 5 | 513 | ||
AK025289.1 | - | 2 | 130 | |||
AK294103.1 | - | 5 | 535 | |||
AK315372.1 | - | 5 | 509 | |||
AL541403.3 | - | 5 | 664 | |||
BC022503.1 | - | 5 | 646 | 4791972 | ||
BC036468.2 | - | 5 | 605 | 4799622 | ||
BX647776.1 | - | 4 | 418 |