Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145036.1 | NP_001138508.1 | 3, 4 | 453 | ||
NM_007357.2 | NP_031383.1 | 3, 4 | 453 | |||
GenBank mRNA | AK295251.1 | - | 3, 4 | 460 | ||
AK312678.1 | - | 3, 4 | 369 | |||
AL832190.1 | - | 3, 4 | 420 | |||
BC014960.1 | - | 3, 4 | 416 | 4876271 | ||
BC051906.1 | - | 3, 4 | 342 | 4541036 | ||
DC349417.1 | - | 3, 4 | 452 | |||
Z34975.1 | - | 3, 4 | 397 |