Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278731.1 | NP_001265660.1 | 3 | 513 | ||
NM_012208.3 | NP_036340.1 | 4 | 588 | |||
XM_011537619.1 | XP_011535921.1 | 5 | 601 | |||
XM_011537620.1 | XP_011535922.1 | 5 | 601 | |||
XM_017009287.1 | XP_016864776.1 | 4 | 588 | |||
XM_017009288.1 | XP_016864777.1 | 5 | 652 | |||
XM_017009289.1 | XP_016864778.1 | 5 | 645 | |||
XM_017009290.1 | XP_016864779.1 | 4 | 638 | |||
XM_017009291.1 | XP_016864780.1 | 4 | 331 | |||
XM_017009292.1 | XP_016864781.1 | 4 | 275 | |||
GenBank mRNA | AK293390.1 | - | 3 | 330 | ||
AK299971.1 | - | 2 | 137 | |||
AK309832.1 | - | 4 | 686 | |||
AK311277.1 | - | 4 | 684 | |||
AK312976.1 | - | 4 | 406 | |||
BC007680.2 | - | 4 | 407 | 3628006 | ||
BC014982.1 | - | 4 | 552 | 4859849 | ||
U18937.1 | - | 4 | 514 |