Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF346 SIRNA Assays ›
Gene Symbol:
ZNF346
Gene Name:
zinc finger protein 346
Gene Aliases:
JAZ, Zfp346
Chromosome Location:
Chr.5: 177022680 - 177081189 on Build GRCh38
UniGene ID:
Hs.484259
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136471
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF346
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308213.1
NP_001295142.1
5
640
NM_001308214.1
NP_001295143.1
3
469
NM_001308215.1
NP_001295144.1
4
565
NM_012279.3
NP_036411.1
4
565
XM_005265860.2
XP_005265917.1
5
585
XM_005265864.3
XP_005265921.1
5
585
XM_011534493.1
XP_011532795.1
5
585
XM_011534494.1
XP_011532796.1
5
585
XM_011534495.2
XP_011532797.1
5
585
XM_011534496.2
XP_011532798.1
5
585
XM_017009294.1
XP_016864783.1
4
565
XM_017009295.1
XP_016864784.1
4
565
XM_017009296.1
XP_016864785.1
4
565
XM_017009297.1
XP_016864786.1
4
565
XM_017009298.1
XP_016864787.1
5
585
XM_017009299.1
XP_016864788.1
4
565
XM_017009300.1
XP_016864789.1
4
855
XM_017009301.1
XP_016864790.1
4
565
XM_017009302.1
XP_016864791.1
6
672
XM_017009303.1
XP_016864792.1
2
628
GenBank mRNA
AF083340.1
-
4
538
AK026627.1
-
4
533
AK131469.1
-
5
585
AK295549.1
-
3
445
AL359576.1
-
4
513
BC007775.2
-
4
520
4301820
BU860103.1
-
4
540
CR749692.1
-
4
513
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of apoptotic process
intrinsic apoptotic signaling pathway by p53 class mediator
p53 binding
double-stranded RNA binding
protein binding
zinc ion binding
enzyme binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0