Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPRE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303050.1 | NP_001289979.1 | 4 | 612 | ||
NM_012326.3 | NP_036458.2 | 4 | 813 | |||
XM_006711967.3 | XP_006712030.1 | 4 | 795 | |||
XM_011532700.1 | XP_011531002.1 | 4 | 588 | |||
XM_017003597.1 | XP_016859086.1 | 4 | 545 | |||
GenBank mRNA | AB025186.1 | - | 4 | 527 | ||
AK312342.1 | - | 4 | 547 | |||
BC011557.1 | - | 4 | 512 | 4561018 | ||
BM560331.1 | - | 4 | 581 | |||
CR536523.1 | - | 3 | 374 | |||
CR541845.1 | - | 3 | 374 |