Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012341.2 | NP_036473.2 | 2 | 161 | ||
GenBank mRNA | AF120334.1 | - | 2 | 113 | ||
AF325353.1 | - | 2 | 90 | |||
AK001548.1 | - | 2 | 108 | |||
AK001552.1 | - | 2 | 163 | |||
AK097093.1 | - | 2 | 108 | |||
AK222861.1 | - | 2 | 108 | |||
AK295231.1 | - | 2 | 108 | |||
AK301721.1 | - | 2 | 276 | |||
AK302219.1 | - | 2 | 70 | |||
BC033784.1 | - | 2 | 84 | |||
BC038975.2 | - | 2 | 122 | 5534986 |