Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNPO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191028.2 | NP_001177957.2 | 8 | 1482 | ||
NM_012470.3 | NP_036602.1 | 8 | 1482 | |||
NR_034053.2 | 9 | 1643 | ||||
XM_011515989.2 | XP_011514291.1 | 8 | 1084 | |||
GenBank mRNA | AF145029.1 | - | 8 | 1192 | ||
AJ133769.1 | - | 8 | 1079 | |||
AK001263.1 | - | 1 | 37 | |||
AK312580.1 | - | 8 | 1209 | |||
BC068530.1 | - | 8 | 1210 |