Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAZ2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300905.1 | NP_001287834.1 | 29 | 8395 | ||
NM_013449.3 | NP_038477.2 | 29 | 8386 | |||
GenBank mRNA | AB002312.2 | - | 29 | 7228 | ||
AK127775.1 | - | 10 | 4138 | |||
CR749379.1 | - | 29 | 8388 |
Gene Symbol: RBMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CF890834.1 | - | 1 | 71 |