Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NCOA6 SIRNA Assays ›
Gene Symbol:
NCOA6
Gene Name:
nuclear receptor coactivator 6
Gene Aliases:
AIB3, ASC2, NRC, PRIP, RAP250, TRBP
Chromosome Location:
Chr.20: 34714774 - 34825649 on Build GRCh38
UniGene ID:
Hs.368971
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136563
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NCOA6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242539.2
NP_001229468.1
3
488
NM_001318240.1
NP_001305169.1
6
3010
NM_014071.4
NP_054790.2
5
780
XM_006723755.3
XP_006723818.1
5
708
XM_011528721.2
XP_011527023.1
5
710
XM_011528726.2
XP_011527028.1
5
710
XM_011528727.2
XP_011527029.1
5
710
XM_017027739.1
XP_016883228.1
5
710
XM_017027740.1
XP_016883229.1
6
829
XM_017027741.1
XP_016883230.1
5
710
XM_017027742.1
XP_016883231.1
5
710
XM_017027743.1
XP_016883232.1
5
710
XM_017027744.1
XP_016883233.1
5
707
XM_017027745.1
XP_016883234.1
5
710
XM_017027746.1
XP_016883235.1
5
710
XM_017027747.1
XP_016883236.1
1
14
XM_017027748.1
XP_016883237.1
5
710
XM_017027749.1
XP_016883238.1
5
710
XM_017027750.1
XP_016883239.1
5
710
XM_017027751.1
XP_016883240.1
5
708
GenBank mRNA
AF128458.1
-
4
641
AF171667.1
-
4
583
AF177388.1
-
4
599
AF208227.1
-
6
3007
AF245115.1
-
3
498
BC114613.1
-
4
596
BC136272.1
-
4
589
9020259
D80003.2
-
4
576
JF707637.1
-
3
439
JF707638.1
-
3
439
JF707639.1
-
3
439
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001242558, NM_019825, BC031113, AK171864, BC083187, and others.
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle development
DNA replication
DNA repair
DNA recombination
DNA-templated transcription, initiation
transcription initiation from RNA polymerase II promoter
cellular response to DNA damage stimulus
brain development
heart development
response to hormone
myeloid cell differentiation
intracellular estrogen receptor signaling pathway
positive regulation of insulin secretion involved in cellular response to glucose stimulus
glucocorticoid receptor signaling pathway
cellular lipid metabolic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
chromatin binding
transcription coactivator activity
protein binding
ligand-dependent nuclear receptor binding
enzyme binding
protein domain specific binding
estrogen receptor binding
ligand-dependent nuclear receptor transcription coactivator activity
glucocorticoid receptor binding
protein homodimerization activity
peroxisome proliferator activated receptor binding
retinoid X receptor binding
thyroid hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-k9xzd:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0