Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270520.1 | NP_001257449.1 | 6, 7 | 912 | ||
NM_014992.2 | NP_055807.1 | 5, 6 | 806 | |||
XM_005267430.1 | XP_005267487.1 | 6, 7 | 964 | |||
XM_005267431.1 | XP_005267488.1 | 6, 7 | 1064 | |||
GenBank mRNA | AB014566.1 | - | 5, 6 | 792 | ||
AK291790.1 | - | 6, 7 | 881 | |||
BC038428.1 | - | 6, 7 | 904 | 4993504 | ||
BC064999.1 | - | 6, 7 | 895 | 5784628 | ||
BX247986.1 | - | 6, 7 | 989 |