Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015129.5 | NP_055944.2 | 3 | 561 | ||
NM_145799.3 | NP_665798.1 | 3 | 561 | |||
NM_145800.3 | NP_665799.1 | 3 | 561 | |||
NM_145802.3 | NP_665801.1 | 3 | 561 | |||
XM_005262400.3 | XP_005262457.1 | 3 | 369 | |||
XM_006724748.2 | XP_006724811.1 | 3 | 369 | |||
XM_006724750.2 | XP_006724813.1 | 3 | 369 | |||
XM_011531317.2 | XP_011529619.1 | 3 | 369 | |||
XM_011531318.2 | XP_011529620.1 | 3 | 369 | |||
GenBank mRNA | AF397023.1 | - | 3 | 406 | ||
AF403058.1 | - | 3 | 561 | |||
AF403059.1 | - | 3 | 561 | |||
AF403060.1 | - | 3 | 561 | |||
AF403061.1 | - | 3 | 561 | |||
AF403062.1 | - | 3 | 561 | |||
AK026589.1 | - | 3 | 354 | |||
AK290977.1 | - | 3 | 613 | |||
AK298487.1 | - | 4 | 468 | |||
AK299273.1 | - | 3 | 416 | |||
AK303116.1 | - | 4 | 438 | |||
AK303224.1 | - | 3 | 484 | |||
AY005981.1 | - | 3 | 453 | |||
AY034177.1 | - | 3 | 551 | |||
AY679521.1 | - | 3 | 561 | |||
BC009291.2 | - | 3 | 345 | 4111673 | ||
BC011922.2 | - | 3 | 349 | 4542179 | ||
BC036240.1 | - | 3 | 388 | 5298284 | ||
BC069231.1 | - | 3 | 333 | 4852319 | ||
D50918.1 | - | 289 |