Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ADAM20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003814.4 | NP_003805.3 | 2 | 721 | ||
XM_005268151.3 | XP_005268208.1 | 2 | 9617 | |||
GenBank mRNA | BC025378.1 | - | 2 | 724 | 4827383 |