Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSTF2T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015235.2 | NP_056050.1 | 1 | 1657 | ||
GenBank mRNA | AB014589.1 | - | 1 | 1405 | ||
AK222574.1 | - | 1 | 1645 | |||
AK314318.1 | - | 1 | 1657 | |||
AL136747.1 | - | 1 | 1653 | |||
AY130299.1 | - | 1 | 1649 | |||
BC028239.1 | - | 1 | 1641 | 5244025 |