Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286715.1 | NP_001273644.1 | 20 | 3068 | ||
NM_001286717.1 | NP_001273646.1 | 20 | 2948 | |||
NM_015239.2 | NP_056054.2 | 21 | 3013 | |||
XM_005251847.3 | XP_005251904.1 | 21 | 3079 | |||
XM_005251848.2 | XP_005251905.1 | 21 | 3257 | |||
XM_005251849.3 | XP_005251906.1 | 20 | 3003 | |||
XM_011518418.2 | XP_011516720.1 | 21 | 3079 | |||
XM_011518420.2 | XP_011516722.1 | 11 | 6332 | |||
XM_017014544.1 | XP_016870033.1 | 21 | 3624 | |||
XM_017014545.1 | XP_016870034.1 | 21 | 2959 | |||
XM_017014546.1 | XP_016870035.1 | 20 | 2846 | |||
XM_017014547.1 | XP_016870036.1 | 20 | 3003 | |||
XM_017014548.1 | XP_016870037.1 | 20 | 2946 | |||
XM_017014549.1 | XP_016870038.1 | 19 | 2965 | |||
GenBank mRNA | AB028958.2 | - | 21 | 2991 | ||
AK001544.1 | - | 22 | 2813 | |||
AK022562.1 | - | 9 | 1656 | |||
AK023280.1 | - | 9 | 1648 | |||
AK024698.1 | - | 5 | 598 | |||
AK295774.1 | - | 20 | 2948 | |||
AK299506.1 | - | 17 | 2508 | |||
AL833359.1 | - | 9 | 1605 | |||
BC060815.1 | - | 21 | 2978 | 30344122 | ||
BX648366.1 | - | 9 | 2060 |