Gene Symbol: UBR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184801.1 | NP_001171730.1 | 2 | 426 | ||
NM_015255.2 | NP_056070.1 | 2 | 426 | |||
XM_005248965.4 | XP_005249022.1 | 2 | 543 | |||
XM_011514438.2 | XP_011512740.2 | 2 | 626 | |||
XM_017010594.1 | XP_016866083.1 | 2 | 627 | |||
XM_017010595.1 | XP_016866084.1 | 2 | 627 | |||
XM_017010596.1 | XP_016866085.1 | 2 | 461 | |||
XM_017010597.1 | XP_016866086.1 | 2 | 628 | |||
GenBank mRNA | AY061884.1 | - | 2 | 128 | ||
BC024217.2 | - | 2 | 405 | |||
BC064512.1 | - | 2 | 405 | |||
DR003057.1 | - | 2 | 433 |