Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NEDD4L SIRNA Assays ›
Gene Symbol:
NEDD4L
Gene Name:
neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase
Gene Aliases:
NEDD4-2, NEDD4.2, RSP5, hNEDD4-2
Chromosome Location:
Chr.18: 58044362 - 58401540 on Build GRCh38
UniGene ID:
Hs.185677
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136862
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NEDD4L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144964.1
NP_001138436.1
25, 26
2659
NM_001144965.1
NP_001138437.1
25, 26
2611
NM_001144966.2
NP_001138438.1
24, 25
2536
NM_001144967.2
NP_001138439.1
25, 26
2705
NM_001144968.1
NP_001138440.1
25, 26
2601
NM_001144969.1
NP_001138441.1
24, 25
2541
NM_001144970.2
NP_001138442.1
23, 24
2584
NM_001144971.1
NP_001138443.1
25, 26
2629
NM_001243960.1
NP_001230889.1
23, 24
2513
NM_015277.5
NP_056092.2
24, 25
2645
XM_005266658.4
XP_005266715.2
22, 23
2636
XM_005266660.4
XP_005266717.2
21, 22
2580
XM_005266663.4
XP_005266720.2
20, 21
2520
XM_006722421.3
XP_006722484.2
21, 22
2575
XM_006722424.3
XP_006722487.2
18, 19
2128
XM_006722425.3
XP_006722488.2
17, 18
2072
XM_006722426.3
XP_006722489.1
27, 28
2853
XM_006722428.3
XP_006722491.1
26, 27
2775
XM_006722430.3
XP_006722493.1
25, 26
2683
XM_011525887.2
XP_011524189.1
27, 28
2833
XM_017025676.1
XP_016881165.1
19, 20
2325
XM_017025677.1
XP_016881166.1
18, 19
2269
XM_017025678.1
XP_016881167.1
25, 26
2714
XM_017025679.1
XP_016881168.1
24, 25
2629
XM_017025680.1
XP_016881169.1
21, 22
2317
XM_017025681.1
XP_016881170.1
20, 21
2119
GenBank mRNA
AB007899.1
-
23, 24
2484
AB071179.1
-
24, 25
2476
AF210730.1
-
24, 25
2436
AF385931.1
-
25, 26
2660
AK294963.1
-
24, 25
2536
AK300444.1
-
24, 25
2487
AL137469.1
-
20, 21
1957
AY112983.1
-
25, 26
2611
AY112984.2
-
25, 26
2601
AY112985.2
-
24, 25
2545
AY312514.1
-
25, 26
2422
BC000621.2
-
19, 20
2062
BC019345.1
-
19, 20
2062
BC032597.1
-
23, 24
2540
5528964
BQ215431.1
-
4, 5
269
DQ181796.1
-
25, 26
2629
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
protein polyubiquitination
regulation of membrane depolarization
protein monoubiquitination
sodium ion transport
cellular sodium ion homeostasis
excretion
response to metal ion
protein ubiquitination
viral life cycle
water homeostasis
ion transmembrane transport
regulation of ion transmembrane transport
regulation of protein catabolic process
regulation of membrane potential
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of protein catabolic process
positive regulation of endocytosis
regulation of membrane repolarization
protein K48-linked ubiquitination
ventricular cardiac muscle cell action potential
regulation of potassium ion transmembrane transporter activity
negative regulation of potassium ion transmembrane transporter activity
negative regulation of potassium ion transmembrane transport
negative regulation of sodium ion transmembrane transport
positive regulation of dendrite extension
negative regulation of protein localization to cell surface
negative regulation of sodium ion transmembrane transporter activity
positive regulation of caveolin-mediated endocytosis
ubiquitin-protein transferase activity
protein binding
potassium channel regulator activity
ligase activity
sodium channel regulator activity
potassium channel inhibitor activity
sodium channel inhibitor activity
ion channel binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-vrvhj:80/10.239.161.228:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0