Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: VPS39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301138.1 | NP_001288067.1 | 10 | 940 | ||
NM_015289.3 | NP_056104.2 | 9 | 907 | |||
XM_011521403.2 | XP_011519705.1 | 10 | 909 | |||
XM_011521404.2 | XP_011519706.1 | 9 | 912 | |||
XM_017022036.1 | XP_016877525.1 | 10 | 908 | |||
XM_017022037.1 | XP_016877526.1 | 9 | 912 | |||
XM_017022038.1 | XP_016877527.1 | 9 | 1095 | |||
XM_017022039.1 | XP_016877528.1 | 9 | 880 | |||
GenBank mRNA | AB018313.2 | - | 9 | 858 | ||
AF280814.1 | - | 10 | 767 | |||
AF281052.1 | - | 9 | 877 | |||
BC015817.2 | - | 5 | 467 | |||
BC068559.1 | - | 9 | 876 | 5295166 | ||
BX537404.1 | - | 9 | 886 |