Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: VPS39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301138.1 | NP_001288067.1 | 26 | 4832 | ||
NM_015289.3 | NP_056104.2 | 25 | 4799 | |||
XM_011521403.2 | XP_011519705.1 | 26 | 4871 | |||
XM_011521404.2 | XP_011519706.1 | 25 | 4874 | |||
XM_017022038.1 | XP_016877527.1 | 25 | 4987 | |||
XM_017022039.1 | XP_016877528.1 | 25 | 4798 | |||
GenBank mRNA | AB018313.2 | - | 25 | 4750 | ||
AF281052.1 | - | 25 | 4769 | |||
AK025334.1 | - | 10 | 2998 | |||
AK074193.1 | - | 1 | 1291 | |||
AK124255.1 | - | 1 | 2792 | |||
AK128403.1 | - | 8 | 4121 | |||
AL117456.1 | - | 1 | 1886 | |||
BC015817.2 | - | 21 | 4359 | |||
BX537404.1 | - | 25 | 4778 |