Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003679.4 | NP_003670.2 | 15 | 2760 | ||
GenBank mRNA | AF056032.1 | - | 15 | 2503 |
Gene Symbol: OPN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM679966.1 | - | 1 | 35 |