Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KHSRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003685.2 | NP_003676.2 | 6 | 605 | ||
XM_005259668.4 | XP_005259725.1 | 6 | 495 | |||
XM_011528395.2 | XP_011526697.1 | 7 | 460 | |||
XM_017027408.1 | XP_016882897.1 | 7 | 459 | |||
XM_017027409.1 | XP_016882898.1 | 7 | 459 | |||
XM_017027410.1 | XP_016882899.1 | 6 | 354 | |||
GenBank mRNA | AK300957.1 | - | 7 | 473 | ||
BC085004.1 | - | 6 | 569 | 6597976 | ||
DC388502.1 | - | 7 | 473 | |||
U69126.1 | - | 6 | 328 | |||
U94832.1 | - | 6 | 588 |